Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard Labels
Small labels serving as visual reminders of biohazards
Specifications
Certifications/Compliance | OSHA 29 CFR 1910.145 |
---|---|
Color | Orange |
Format | Label |
Material | Paper |
For Use With (Application) | Biohazard |
Catalog Number | Mfr. No. | Display Text | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Display Text | Price | Quantity & Availability | |||||
17-986-90D
|
National Marker™
WOL 5 |
Biohazard with pictogram, Hazard Identity with space to write-on |
Item Discontinued
This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
17-986-90F
|
National Marker™
WOL 7 |
Biohazard with pictogram |
Item Discontinued
This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
Description
- Material: Pressure-sensitive paper
- Size: 2.54 × 7.62cm (1 × 3 in.)
- Color: Orange with black imprint
Specifications
OSHA 29 CFR 1910.145 | |
Label | |
Biohazard | |
1 x 3 in. |
Orange | |
Paper | |
1 Roll | |
2.54 x 7.62 cm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title