Learn More
Proteintech Group Inc Nano-Secondary alpaca anti-rabbit IgG, recombinant VHH, for 2x Cys conjugation [CTK0102]
SDP
Supplier:  Proteintech Group Inc SRBGCYS22500
Encompass
Monovalent, recombinant secondary single domain antibody to rabbit IgG: alpaca monoclonal Nanobody, Fab-specific, for cysteine conjugation with thiol-reactive reagents, e.g. maleimides
Catalog No. 50-225-8861
Explore available promotions