Learn More
Proteintech Group Inc Nano-Secondary alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH, Alexa Fluor 568 [CTK0101, CTK0102]
SDP
Supplier:  Proteintech Group Inc SRBAF568110
Encompass
Nano-Secondary alpaca anti-human IgG/anti-rabbit IgG, recombinant VHH is an anti-human IgG and anti-rabbit IgG specific secondary antibody. It consists of of a mixture of 2 Nanobodies that bind to human IgG and rabbit IgG with high affinity & specificity.
Catalog No. 50-225-8856
Explore available promotions