Learn More
Aat Bioquest MUG [4-Methylumbelliferyl-beta-D-galactopyranoside] CAS 6160-78-7 (10 g)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Aat Bioquest 14012
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
4-Methylumbelliferyl-beta-D-galactopyranoside MUG is a fluorogenic MU substrate for detecting galactosidase in cell extracts and purified enzyme preparations using a fluorescence microplate reader or fluorometer. This catalog is for 10 g.
This item is not returnable.
View return policy