Learn More
MTC Bio TotalPure Metal Free Centrifuge Tubes 15mL Individually Wrapped
Supplier: MTC Bio C2915W
Total-Pure™ Centrifuge Tubes are produced from a proprietary purified polypropylene blend that is certified to be free of trace metal content. Tested via ICP-MS, these tubes are certified to contain less than 1ppm of common trace metals including: Calcium, Magnesium, Zinc, Iron, Manganese, Copper, Aluminum, Silicon, Nickel, Vanadium, Sodium, Phosphorous, Cobalt, Chromium, Potassium, Lithium, Lead, Selenium, Cadmium, Mercury, and Arsenic.In addition to a superior level of purity, TotalPure™ Tubes include a wide array of premium features. These tubes also feature Hi-Viz™ graduations that are easily identified at any angle and the printed max-freeze line indicates the maximum allowable volume to be frozen inside the tubes. TotalPure's natural color caps offer a smooth seal that is easy to open/close with one hand without compromising the integrity of the 95kPA leak-proof seal.• Ultra-Pure Polypropylene Blend Certified Free of Trace Metals
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.