Learn More
MSA™ V-Gard™ General Purpose Visor
Polycarbonate visor with anti-fog/anti-scratch coatings
Supplier: MSA™ 10115841
Description
- Contoured visor with closer fit and resistance to impact and splash hazards
- Nitrometer for use with or without V-Gard Retractable or Standard Chin Protectors
- Clear AF/AS coated visor for industrial impact applications
- Offers maximum UV protection (“U6”)
- Size: 20.3 x 43.2 x 0.152cm (8 x 17 x 0.06 in.)
For hot/humid conditions, chipping, splash, chiseling, drilling, grinding, machining, power fastening, sanding and sawing
Certifications
Impact-rated (Z87+),ANSI/ISEA Z87.1-2010 and CSA Z94.3, Third-party by Intertek and CSA International