Learn More
MSA™ PremAire™ Cadet Respirator Kit Accessories: Air-Line Respirator
Accessory for PremAire Cadet Respirator Kits
Supplier: MSA™ 10108495
Includes: Push-To-Connect Firehawk Regulator, nylon belt assembly, Snap-Tite Quick-Disconnect and low-pressure TransportAire Portable Air Supply with 30-minute carbon fiber cylinder and eight-foot coiled nylon air supply hose
Description
For use with PremAire Cadet Respirator Kits (Cat. Nos. 17-200-023 and 17-200-025).
- Type C respirator supplies on-demand respiratory protection
- Mask-mounted regulator
Specifications
Supplied Air Respirator Assembly | |
PremAire Cadet Respirator Kits | |
PremAire Cadet Respirator with Advantage 4000 Facepiece (medium size, hycar rubber, polyester net head harness) with Push-To-Connect Firehawk Regulator, nylon belt assembly, Snap-Tite Quick-Disconnect and low-pressure TransportAire Portable Air Supply with 30-minute carbon fiber cylinder, eight-foot coiled nylon air supply hose | |
10108495 |