Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSA™ Military Style CBRN and Riot Control Gas Mask Pouch
Storage and carrying bag for use with gas masks.
Supplier: MSA™ 10034184
Description
- For use with gas masks
- Pouch, Black
- 15.5 x 13 x 4.5 inches
Specifications
Gas Mask Pouch | |
10034184 | |
CBRN, Civil Disturbances, Clandestine Labs | |
NIOSH CBRN |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction