Learn More
Waters Corp MS Reference Standard/ Horse Heart Myoglobin
Supplier: Waters Corp 700004749
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Mass Spectrometry standard for system assessment. 2 X Horse Heart Myoglobin 2.5 mg ±0.125 mg 15 mL Clear Nalgene Bottle 1 X 715002043 Sample Preparation Document Certificate of Analysis