Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Brewers Yeast
Used as an insect diet.
Supplier: MP Biomedicals™ 0290331225
Description
- Source of B-complex vitamins, chromium and selenium
- Brewers Yeast is commonly used for baking break, fermenting wine and for beer and ethanol production
Specifications
Animal Diet | |
Source of B-complex Vitamins, Chromium and Selenium, also used to bake bread, ferment wine, beer and Ethanol production | |
S. cerevisiae |
Powder | |
11.33 kg | |
15°C to 30°C |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction