Learn More
Kent Scientific MouseSTAT™ Pulse Oximeter Whole Body Sensor
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For use with PhysioSuite and PhysioSuite Modules.
$415.20 - $692.66
Specifications
Animal | Mouse and pups that are greater than 3 days old |
---|---|
For Use With (Application) | Vital Sign Monitoring, Pulse Oximetry, Heart Rate, Physiological Monitoring |
For Use With (Equipment) | PhysioSuite and PhysioSuite Modules |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
13-005-137
![]() ![]() |
Kent Scientific
SPO213MM |
Pulse Oximeter Whole Body Sensor |
Each for $415.20
|
|
|||||
13-005-138
![]() ![]() |
Kent Scientific
SPO213MMMRI |
Pulse Oximeter Whole Body Sensor, MRI Compatible |
Each for $692.66
|
|
|||||
Description
- Recommended for use with mouse and pups that are greater than 3 days old
- MRI compatible sensor includes 3m cable
Applications
Vital sign monitoring, pulse oximetry, heart rate, physiological monitoring
Specifications
Mouse and pups that are greater than 3 days old | |
Vital Sign Monitoring, Pulse Oximetry, Heart Rate, Physiological Monitoring | |
PhysioSuite and PhysioSuite Modules |