Learn More
Motic Digital Pathology MoticEasyScan Pro 6 Digital Slide Scanner
The MoticEasyScan Pro is a gateway product for any lab, startup, or medical organization joining the digital revolution in pathology. It scans and uploads high-quality digital slides to your slide management server, all at a single click.
Supplier: Motic Digital Pathology FSCMDPPKG2
Description
- Simple and easy to use, this scanner system offers 1-slide and 6-slide capacity options.
- With its small footprint, high resolution power, and hassle-free setup, this multipurpose scanner is the ideal tool for any application that requires glass slide digitization.
- MoticEasyScan scanners use a specially designed high-NA Plan Apochromatic objective (20X/0.75) to maximize color fidelity and resolution power.
- Combined with a large 2/3 in. CCD sensor, the MoticEasyScan is able to image large fields of view in a split second, while delivering image detail equivalent to a high power 40X lens.
Specifications
Slide Scanner | |
33 x 33 x 44 cm | |
MoticEasyScan |
Scanning Camera 12 MP | |
Any Digital Slide Application | |
MoticEasyScan Pro 6 Single Objective Base, Computer for MoticEasyScan Pro 6, Online Customer Training |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov