Learn More
Motic Digital Pathology MoticEasyScan One Digital Slide Scanner
The MoticEasyScan One is compact, powerful, and deployable anywhere. This scanner is the perfect assistant for any digital slide application. Share your slides with a single click, or switch to live mode for direct specimen observation.
Supplier: Motic Digital Pathology FSCMDPPKG1
Description
- This scanning system sets a new industry benchmark with its affordable price and high performance, seamlessly integrating into any workflow and bringing lab-quality scanning to any work environment.
- MoticEasyScan scanners use a specially designed high-NA Plan Apochromatic objective (20X/0.75) to maximize color fidelity and resolution power.
- Combined with a large 2/3 in. CCD sensor, the MoticEasyScan is able to image large fields of view in a split second, while delivering image detail equivalent to a high power 40X lens.
Specifications
Slide Scanner | |
20.5 x 39.8 x 42 cm | |
MoticEasyScan |
Scanning Camera 5 MP | |
Any Digital Slide Application | |
MoticEasyScan One Single Bbjective Base, Computer for MoticEasyScan One, Online Customer training |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov