Learn More
Maine Molecular Quality Controls Inc FilmArray BCID2 Control Panel M416
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Maine Molecular Quality Controls Inc M416
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The FilmArray BCID2 Control Panel M416 is intended for use as an external positive and negative assayed quality control to monitor the performance of in vitro laboratory nucleic acid testing procedures for the qualitative detection of antimicrobial resistance genes, gram positive bacteria, gram negative bacteria, and yeast pathogens on the BioFire Blood Culture Identification 2 (BCID2) Panel on the BioFire FilmArray systems. FilmArray BCID2 Control Panel M416 is comprised of 2 controls, FilmArray BCID2 Positive Control and FilmArray BCID2 Negative Control. FilmArray BCID2 Control Panel M416 is comprised of 12 tubes, 6 tubes of positive control and 6 tubes of negative control, 200μL each.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.