Learn More
Electron Microscopy Sciences Miniature Measuring Scale #3
Supplier: Electron Microscopy Sciences 6213503
These unit measuring scales may be used directly under a microscope or magnifier for quick measuring of small specimens, parts, grooves, slots, line widths, holes, chambers, etc. Precision fabricated from stainless steel, the scales are welded to bendable stainless steel shafts and mounted in color coded, non-rolling handles. Each scale is individually packed in a plastic tube. Overall length of the scale is 83 mm (3 1/4"), including handle. Calibrated to 0.1mm, 10mm total
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.