Learn More
United Biosystems Inc MINI CELL SCRAPERS 200/PK
Supplier: United Biosystems Inc MCS200
Effectively harvest cells or cell lysates from 96-, 48- and 24-well plates Size: 3/8" (0.5 cm) in width and 2 3/8" (6 cm) in length Polyethylene, disposable & sterile