Learn More
MilliporeSigma Milli-Q™ SQ 240 Water Purification System Kit
The Milli-Q™ SQ 240 Purification System Kit is composed of the Milli-Q™ SR 240 and the Milli-Q™ SQ 200 water purification systems. Delivers pure and ultrapure water from tap water source.
Supplier:  MilliporeSigma ZSQ240R0TK
Catalog No. ZSQ240R0TK
Explore available promotions
Add to Cart
Description
Description
The Milli-Q™ SQ 240 Purification System Kit is composed of the Milli-Q™ SR 240 and the Milli-Q™ SQ 200 water purification systems. The Milli-Q™ SR 240 water purification system delivers pure reverse osmosis (RO) water to a mobile SQ Switch 3.5 L tank. It can be equipped with a Gravity Valve (optional) to dispense pure RO water directly from the SQ Switch tank base. The Milli-Q™ SQ 200 water purification system delivers ultrapure (Type 1) water from the SQ Switch 3.5 L tank filled with pure RO water produced by the Milli-Q™ SR 240 system. It benefits from a flexible handset dispenser to deliver ultrapure water. Kit requires 2x Power Cords, Cartridges, Final Filter, and Installation (not included).
Features and Benefits
  • Reliable 18.2 MΩcm resistivity ultrapure (Type 1) water quality is indicated at a glance by a colored LED
  • TOC is reduced by an integrated photo-oxidation UV lamp
  • Freshly dispensed ultrapure water is available anywhere, in or out of the lab, from the autonomous Milli-Q™ SQ 200 ultrapure dispensing module thanks to the mobile SQ Switch tank
  • Fast (as little as 6 min) refilling of the mobile SQ Switch tank with freshly produced pure RO water due to the high-flow RO cartridge (up to 42 L/h) of the pure water production station; RO water recovery loop saves 50% of the rejected water; automatic RO membrane rinsing maintains its high performance.
  • Compact setup and self-guided installation manual and videos let you plug the system wherever is convenient for your lab activity and move it later if needed
  • Unlimited scalability with one pure RO water production station able to feed multiple autonomous Milli-Q™ SQ 200 ultrapure dispensing modules (just plug in an electrical outlet); Supports lab activity expansion while limiting budget investment
  • Individual Twist and Lock cartridge replacement simplifies maintenance and limits running cost
  • Compact size plus water and energy savings support lab sustainability efforts
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrated
Type 1 and 3
Water Purification System
Potable Tap
Yes
100/240 V
Product Suggestions
Product Suggestions