Learn More
PRECIGENOME MICROFLUIDIC MIXER MIX-4 CHIP
Supplier: PRECIGENOME CHPMIX4
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Microfluidic Mixer MIX-4 Chip for NanoParticle Synthesis Microfluidic Mixer Chip, which is designed for NanoParticle Synthesis.4 channels (devices)/pcChip Type: Micro-mixer Connection: Female Luer
This item is not returnable.
View return policy