Learn More
Microbiologics™ EZ-Accu Shot™ Microorganism Preparations
For easy growth promotion testing of culture media
$696.00
Specifications
Form | Pellets |
---|---|
Product Type | EZ-Accu Shot |
Includes: Product warranty and peel-off Certificate of Assay.
Catalog Number | Mfr. No. | ATCC® | Species | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | ATCC® | Species | Price | Quantity & Availability | ||||
23-004-133
|
Microbiologics™
0317A |
ATCC 19404 | Clostridium sporogenes |
Each for $696.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-004-130
|
Microbiologics™
0392A |
ATCC 16404 | Aspergillus brasiliensis |
Each for $696.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-004-132
|
Microbiologics™
0443A |
ATCC 10231 | Candida albicans |
Each for $696.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-004-135
|
Microbiologics™
0483A |
8739™* | Escherichia coli |
Each for $696.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-004-136
|
Microbiologics™
0484A |
9027™* | Pseudomonas aeruginosa |
Each for $696.00
|
|
||||
23-004-137
|
Microbiologics™
0485A |
6538™* | Staphylococcus aureus |
Each for $696.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-004-131
|
Microbiologics™
0486A |
ATCC 6633 | Bacillus subtilis |
Each for $696.00
|
|
||||
23-004-134
|
Microbiologics™
0487A |
11437™* | Clostridium sporogenes |
Each for $696.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-001-908
|
Microbiologics™
0363A |
14028™* | Salmonella enterica |
Each for $696.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Each lyophilized microorganism preparation is designed to deliver less than 100CFU per 0.1mL of hydrated suspension as recommended by the USP/JP/Ph. Eur. With little to no prep time required and up to eight hours of stability, EZ-Accu Shot adds efficiency and flexibility to QC lab schedules. Each 1.2mL vial of hydrated suspension offers 10 inocula. For added convenience, a peel-off Certificate of Assay is included for quality control documentation.
- Instant dissolve, no pre-incubation for minimal prep time
- One pellet per vial eliminates the need for forceps, reducing the risk of contamination
- Eight-hour stability extends time and flexibility to perform tests
- Precise CFU count ensures accurate results
- Easy storage with no dry ice required, reducing shipping costs
- Peel-off Certificate of Assay for complete, convenient documentation
- Online Certificate of Analysis for documentation of phenotypic, macroscopic and microscopic test results
- Traceable to USP reference culture or assured quality
Order Info
Each kit contains five vials of a single enumerated microorganism (one lyophilized pellet per vial) and five vials of hydrating fluid (1.2mL per vial).
Specifications
Pellets | |
EZ-Accu Shot |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.