Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Epower™ Staphylococcus aureus subsp. aureus ATCC™ 6538™†
![SDP](/content/dam/fishersci/glyphs/sdp.png)
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
Specifications
Species | Staphylococcus aureus subsp. aureus |
---|---|
ATCC® | 6538 |
Biosafety Level | 2 |
Form | Lyophilized Pellet |
For Use With (Application) | For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination. |
Catalog Number | Mfr. No. | Mean Assay Value | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Mean Assay Value | Price | Quantity & Availability | |||||
12-111-291
![]() |
Microbiologics™
0485E7CRM |
1.0-9.9 + 7 CFU/pellet |
Each for $927.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
12-111-290
![]() |
Microbiologics™
0485E3CRM |
1.0-9.9 + 3 CFU/pellet |
Each for $927.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Epower
- Lyophilized microorganism preparations
- Biosafety Level: 2
- Pellets can be used individually or combined for a mixed microorganism population challenge
- Easily manipulated to deliver a wide variety of concentrations
- Available in concentrations ranging from 102 to 108 CFU per pellet
- ATCC™ 6538™†
Specifications
Staphylococcus aureus subsp. aureus | |
2 | |
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination. |
6538 | |
Lyophilized Pellet | |
10/Pk. |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title