Learn More
Electron Microscopy Sciences Metallurgical Mount
Supplier: Electron Microscopy Sciences 7595201
Accommodates up to 31.8 mm (1.25") diameter of a metallurgical specimen with at least 0.8 mm (1/8") clearance, pin is 3.1 mm (1/8"), depth is 8 mm (5/16")