Learn More
Bioassay Systems meta-Phosphoric Acid for Glutatione Assay. meta-Phosphoric Acid is a chemical reagent used for sample deproteination with the GSH/GSSG Assay Kit (cat no: EGTT-100).
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems MPA2G
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
meta-Phosphoric Acid is a chemical reagent used for sample deproteination with the GSH/GSSG Assay Kit (cat#: EGTT-100).