Learn More
Medical Packaging Cytology Fixative
Forms a protective coating over specimens for safe storage and transportation of slides
Supplier:  Medical Packaging CF4040B12
Catalog No. 22-037-750
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Applies a clear, water-soluble wax coating fix and protect from smears
  • Ideal for cervical smears, which have a tendency to detach from the slide
  • Won't cloud or obscure specimen
  • Environmentally friendly; contains no CFCs
Specifications
Specifications
Cervical smears, which have a tendency to detach from the slide
4 oz.
118.2 mL
Product Suggestions
Product Suggestions