Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MAS™ UrichemTRAK Controls
Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient vial format.
Catalog Number | Mfr. No. | Description | Level | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Level | Price | Quantity & Availability | ||||
23-139110
|
Thermo Scientific™
UR22002 |
UrichemTRAK Controls, Assayed | 2 | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
23-139-109
|
Thermo Scientific™
UR11001 |
UrichemTRAK Controls, Assayed | 1 | N/A | N/A | ||||
This product is regulated by the federal government, and may not be ordered online.This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance. | |||||||||
Description
Monitor the most commonly tested urine chemistry analytes with liquid, assayed Thermo Scientific™ MAS™ UrichemTRAK Controls, available in a convenient Vista™ vial format. Ready-to-use and requiring no additional preparation, these controls will not interfere with ISEs.
- Liquid, human urine matrix
- Open and use; no equilibration required
- Two-year open-vial stability
- Individual levels or multi-pack configurations
- Stable for 24 months at 2° to 8°C
- Assayed values available for most major analyzers
Specifications
2°C to 8°C | |
Urine | |
Control |
Safety and Handling
ShelfLife : 24 months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title