Learn More
Vetequip Inc MAPLESON E BREATHING CIR.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Vetequip Inc 921412
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item has a minimum qty of 3 per supplier requirements.
Mapleson E Breathing Circuit, Tube-outside-a-tube circuit. The fresh gas tube is outside the larger diameter downtube.