Learn More
Bioassay Systems QuantiChrom™ Magnesium Assay Kit. For quantitative determination of Mg2+ and evaluation of drug effects on magnesium metabolism.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems DIMG250
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative determination of Mg2+ and evaluation of drug effects on magnesium metabolism. Key Features: Sensitive and accurate. Use as little as 5 µL sample. Linear detection range 0.1 mg/dL (41 µM) to 3 mg/dL (1.2 mM) Mg2+ in 96-well plate assay. Simple and high-throughput. The procedure involves addition of two reagents and measuring OD500nm. Can be readily automated as a high-throughput assay for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvet or 96-well plate assay. Low interference in biological samples. Assays can be directly performed in serum and urine samples. Method: OD500nm. Samples: Biological, food and beverage. Species: All. Procedure: Assay takes 10 min. Kit size: 250 tests. Detection limit: 0.1 mg/dL (41 µM).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.