Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mechanix Wear™ M-Pact™ 2 Gloves - Red
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Thermoplastic Rubber (TPR) finger guards and metacarpal protection prevent impact injuries
- Accordion knuckle guard with high density EVA foam padding
- Armortex™ fingertips provide durable gripping power
- D3O™ palm padding absorbs and dissipates high-impact energy in the palm
- TrekDry™ material helps keep hands cool and comfortable
- Extended Neoprene cuff provides added protection
- Hook and Loop closure provides a secure fit
- Machine washable
Specifications
Specifications
Cuff Material | Neoprene |
Color | Red |
Hand | Specific |
Material | Thermoplastic Rubber |
Product Type | Mechanic's Gloves |
Size | 2X-Large |
For Use With (Application) | Construction, Heavy Equipment Operation, Maintenance & Repair, Manufacturing, Search & Rescue, and DIY Home Improvement |
Glove Cuff Style | Hook and Loop |
Series | M-Pact™ 2 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction