Learn More
New England Biolabs, Inc. Luna Universal Probe One-Step RT-qPCR Kit – 200 reactions
SDP
Supplier:  New England Biolabs, Inc. E3006S
Encompass
In the Luna Universal One-Step Probe RT-qPCR Kit, Hot Start Taq DNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55°C. For difficult targets/templates, higher RT step temperatures of up to 60°C can be used without compromising Luna performance. Note that to ensure full activation of the WarmStart Luna RT, incubation at temperatures lower than 50°C is not recommended.
Catalog No. 50-152-6347
Explore available promotions