Learn More
Electron Microscopy Sciences Large Double Slot Set Screw Vise, 25 mm
Supplier: Electron Microscopy Sciences 7595104
SEM specimen holder with two 2.5 mm by 5 mm deep slots. Clamp thin specimens and cross sections without conductive paint or tape. 25 x 8 mm with 3.2 mm pin. Machined aluminum with 2 stainless steel allen set screws. Includes allen wrench.