Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ BioWhittaker™ Medium 199 with Hanks' BSS
Supplier: Lonza™ 12118F
Description
- Uses include virology and vaccine production
- Components include purines (adenine, guanine, xanthine, hypoxanthine), pyrimdines (thymine, uracil) and fat soluble compounds
Specifications
2° to 8°C | |
Sterile | |
7.14 to 7.48 |
M199/HBSS | |
Sterile-filtered through two consecutive 0.2 μm filters | |
500 mL |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction