Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lonza™ AccuGENE™ 1M Tris HCl Buffers
Ready-to-use solutions ideal for a wide range of molecular biology applications
$207.68 - $321.63
Specifications
Concentration | 1 M |
---|---|
DNase | Yes |
For Use With (Application) | Ideal for a wide range of molecular biology applications |
Quantity | 1000 mL |
Description
- Prepared with 18 megohm water
- Filtered using 0.2μ filter and filled into sterile bottles
- Manufactured according to strict quality control standards to ensure consistency
- Guaranteed DNase, RNase and protease-free
- Customized solutions are available to meet individual needs
Specifications
1 M | |
Yes | |
Ideal for a wide range of molecular biology applications | |
1000 mL |
For research or manufacturing use only.
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title