Learn More
Bioassay Systems EnzyFluo™ Human/Mouse LKB1(S431) Phosphorylation ELISA Kit. For determination of LKB1(S431) phosphorylation status in whole cells and evaluation of pathway modulation by activators, inhibitors, siRNA etc.
SDP
Supplier:  Bioassay Systems ELKB1100
Encompass_Preferred
For determination of LKB1(S431) phosphorylation status in whole cells and evaluation of pathway modulation by activators, inhibitors, siRNA etc. Key Features: New and improved. Total assay time reduced from the standard 21 hours to 6.5 hours (hands-on time 2.5 hrs). Simple and convenient. Cells are directly cultured in 96-well plates. No cell lysis necessary. Accurate and high-throughput. Protein phosphorylation is normalized to total cellular protein in the same well, greatly minimizing well-to-well variations. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: Cell-Based ELISA (FL530/585 nm, 360/450 nm). Samples: Cells. Species: human, mouse, and rat. Procedure: Assay takes Assay takes 6.5 hrs, hands-on time 2.5 hrs. Kit size: 100 tests.
Catalog No. 50-225-5884