Learn More
Electron Microscopy Sciences K-kits Liquid TEM Kit, 0.2 µm Gap
Supplier: Electron Microscopy Sciences K7260602
An Innovative Enclosed Specimen Holder for Liquid TEM. K-kits are sample holders designed to facilitate convenient TEM observation of liquid samples, allowing nanoobjects, aggregates, and agglomerates (NOAAs) in liquid samples to be characterized. With vacuum compatible sealing of liquids in electron-transmitting thickness, K-kits are micro reaction chambers for countless experiments in materials, chemical, and biological research.
Gap: 0.2 µm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.