Learn More
Kenlor Industries Inc Kenlor Dropper Urine Control Level 1 - 6 X 10 mL
Supplier: Kenlor Industries Inc 210303
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Kenlor semi-quantitative dipstick liquid urine control has values for Leukocytes, Urobilinogen, Bilirubin, Glucose, Blood, pH, Nitrite, Ketones, Protein, & Specific Gravity, plus HCG, Creatine and Microalbumin. Stable for 2 years at 2-8°C, 30 days at room temperature. Stable for 30 days at room temperature or 180 days at 2-8°C after opening.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.