Learn More
Kenlor Industries Inc Kenlor Dipper Urine Control Level 2 - 10 X 15 mL
Supplier: Kenlor Industries Inc 260304
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Kenlor semi-quantitative dipstick liquid urine control has values for Leukocytes, Urobilinogen, Bilirubin, Glucose, Blood, pH, Nitrite, Ketones, Protein, & Specific Gravity, plus HCG, Creatine, Microalbumin. Stable for 2 years at 2-8°C, 30 days at room temperature. Stable for 30 days at room temperature or 120 days at 2-8°C after opening, or 30 uses.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.