Learn More
Chemglass Life Sciences Reaction Vessel Lid, 4-Neck, 29/42 Center Neck, 2-24/40 Side Necks, 1- #7 Chem-Thread Side Neck, 150mm Schott Flange
Supplier: Chemglass Life Sciences CG194404
Reaction Vessel Lid, 4-Neck, 29/42 Center Neck, 2-24/40 Side Necks, 1- #7 Chem-Thread Side Neck, 150mm Schott Flange. four neck vessel lid to fit CG-1920 through CG-1936 vessels. The #7 Chem-Thread will allow a thermometer or bleed tube having an O.D. of 4 to 7mm to be inserted vertically to the bottom of the vessel. Thread will accept CG-3504 adjustable immersion thermometers. Lid has a flat ground surface allowing it to be greased or used with an o-ring.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.