Learn More
CTL Scientific Supply Corp LEAD ACETATE PAPER,PK/3 DISP.
Supplier: CTL Scientific Supply Corp 907443
LEAD ACETATE PAPER - Pack of 3 dispensers each containing a 5 meter roll of paper. Limit of sensitivity: 1 drop of a solution of 5 mg/liter sulfide gives a brown ring, which is just visible.