Learn More
Monarch Instrument Pocket Laser Tach 200, Digital Tachometer
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Monarch Instrument 6125011
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Kit, Tachometer; Pocket Laser 200; Includes: Tach, RCA, Carrying Case, N.I.S.T. and 5 ft. roll of T-5 reflective tape; Operates up to 25 ft. using a laser light source; Accepts Remote Sensors; Dimensions: 6.92H x 2.4W x 1.6 in. D