Learn More
Fujifilm Healthcare Americas Corp L-Type Triglyceride M Enzyme Color B
Supplier: Fujifilm Healthcare Americas Corp 99002991
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The L-Type Triglyceride M Enzyme Color B (4 X 20 mL) is B is for use in the L-Type Triglyceride M Enzyme assay. The L-Type Triglyceride M Enzyme assay is an in vitro assay for the determination of triglycerides in serum or plasma. Package size: 4 x 20 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.