Learn More
Hach Company HQ440D Laboratory Nitrate (NO₃⁻) Ion Meter Package with ISENO3181 Ion Selective Electrode
Supplier: Hach Company 8508100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Designed for water quality experts, the Hach HQ440D Laboratory Multi-Meter is an advanced laboratory meter that takes the guesswork out of measurements. HQD meters connect with smart Intellical electrodes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced HQD Laboratory Meter is designed to withstand years of continuous lab use. HQD meters feature a simple user interface that does not require manuals or training to operate. The Intellical ISENO3181 is a digital, combination nitrate ion selective electrode (ISE) with a non-refillable, double junction reference and built-in temperature sensor. The Intellical ISENO3181 electrode is intended for laboratory use. The ISENO3181 is ideal for measuring nitrate concentrations in wastewater, drinking water and general water quality applications. The ISENO3181’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.