Learn More
Excel Equipment and Supply Kolor Kut 2.5 OZ Modified Water Finding Paste
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Excel Equipment and Supply KOL25KKM
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Unique Kolor Kut Modified Water Finding Paste works with a measuring gauge, rod, or calibrated line to determine water levels. Apply the paste in the area that the level is expected to appear. When the paste comes in contact with water, it will immediately change from golden brown to bright red, marking the level. Gauges water levels in oxygenated fuels. Paste wipes off easily after use and leaves no residue.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.