Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Koehler™ Instrument Multi-Strip Vise for LPG Copper Strip Corrosion Test Water Bath
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Four sample testing capability
Supplier: Koehler™ Instrument K25090
Description
- For use with LPG Copper Strip Corrosion Test Water Bath
- Thermostatically controlled
- Holds four copper strips at a time, firmly in place without marring edges
- Controls temperature at 37.8 ±0.5°C (100 ±1°F) per ASTM specifications
- Soxhlet reflux condenser and constant water level device maintain proper working depth
- Stainless-steel double-wall construction
- Fully insulated
- Dimensions: 30 × 25 × 61cm (12 × 10 × 24 in.)
Compliance
Conforms to the specifications of ASTM D1838; GPA 2140; ISO 6251
Specifications
LPG Copper strip corrosion test water bath | |
Stainless Steel |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction