Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column with Fritted Disc, PTFE Stopcock Plug and Joint
SDP
Gravity chromatography column
Supplier:  DWK Life Sciences 4205500530
 View more versions of this product
Catalog No. K4205500530
Explore available promotions
Add to Cart
Description
Description
  • Medium wall tubing
  • 40-60 micron pore size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mm
Gravity Chromatography Column
580 mL
Glass
420570 Series reservoir and 420575 series flow control adapters
40 to 60 μm
300 mm
Fritted disc, 2mm bore PTFE stopcock plug and 24/40 standard taper upper joint
Product Suggestions
Product Suggestions