Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A20 Shoe Covers
Click to view available options
White; Universal
Description
- Breathable, patented Microforce™ Barrier SMS Fabric
- Standard sole
- Fully elasticized
- White
Specifications
Specifications
Size | Universal |
Material | SMS |
Color | White |
Closure Type | Fully Elastic |
Includes | Traction |
For Use With (Application) | Laboratories, Medical Facilities |
Certifications/Compliance | NFPA 99 |
Product Type | Shoe Covers |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction