Learn More
Bioassay Systems EnzyChrom™ Ketone Body Assay Kit. For quantitative determination of ketone body (acetoacetate and 3-hydroxybutyrate) and evaluation of drug effects on its metabolism.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bioassay Systems EKBD100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For quantitative determination of ketone body (acetoacetate and 3-hydroxybutyrate) and evaluation of drug effects on its metabolism. Key Features: Sensitive and accurate. Uses 10 µL sample. Linear detection range of 0.12 to 8 mM for each ketone body in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the optical density at room temperature. High-throughput. Can be automated as a high-throughput 96-well plate assay for many samples per day. Method: OD340nm. Samples: Serum, plasma, urine etc. Species: All. Procedure: Assay takes 30 min. Kit size: 200 tests. Detection limit: 0.12 mM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.