Learn More
Mystaire™ Isola™ Vue Filtered Workstation
Keep workers safe and your work environment up to OSHA requirements. Isola™ vue filtered workstations are fitted with the EverSafe™ III TOUCH advanced microprocessor safety controller.
$8411.00 - $11262.00
Catalog Number | Mfr. No. | Width (English) | Width (Metric) | Amperage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Width (English) | Width (Metric) | Amperage | Price | Quantity & Availability | |||
15-338-900
|
Mystaire™
MYISL36 |
36.8 in. | 934.72 mm |
Each for $8,411.00
|
|
||||
15-338-901
|
Mystaire™
MYISL48 |
48 in. | 121.9 cm |
Each for $9,773.00
|
|
||||
15-338-902
|
Mystaire™
MYISL72 |
72.8 in. | 1849.12 mm |
Each for $11,262.00
|
|
||||
Description
- EverSafe™ III TOUCH touch screen controller constantly monitors fume hood performance with audible and visible alarms
- Vividly displayed status of face velocity in linear feet per minute or meters per second
- 18-Icon display for simple touch control of hood features and functions
- LED lighting for true illumination of the entire work area
- Three-stage filtration to confirm to SEFA 9-2010 DHIII recommendations
- Polycarbonate construction for 360 degree visibility— excellent for demonstrations
- Variable airflow control with automatic airflow sensor calibration
- Solid-state gas detection with three sensitivity set points
- Conforms to UL, CSA and CE electrical requirements
- Integral polypropylene spill basin with rolled foil entry
- 110 or 220 VAC
- Built-in filter and application validation
- Optional installation of polypropylene sink
Specifications
28 in. |