Learn More
Bioassay Systems QuantiChrom™ Isocitrate Dehydrogenase Assay Kit. For quantitative colorimetric kinetic determination of isocitrate dehydrogenase activity and evaluation of drug effects on its metabolism.
SDP
Supplier:  Bioassay Systems DIDH100
Encompass_Preferred
For quantitative colorimetric kinetic determination of isocitrate dehydrogenase activity and evaluation of drug effects on its metabolism. Key Features: Fast and sensitive. Linear detection range (20 µL sample): 0.1 to 100 U/L for 30 min reaction. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD565nm. Samples: Plasma, serum,tissue and culture media etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.1 U/L.
Catalog No. 50-225-5863
Explore available promotions