Learn More
World Precision Instrument ISO-OXY-2 or OXELP Sleeve Kit
Supplier: World Precision Instrument 5378
Replacement sleeves for your oxygen sensors
These sleeves fit the ISO-OXY-2 or the OXELP oxygen sensors. These sensors are designed for use in the WPI TBR4100/TBR1025 system.