Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine, Resublimed (DEA List I Chemical), Crystal, USP, 99.8-100.5%, Spectrum™ Chemical
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Click to view available options
Quantity:
25 g
125 g
500 g
6 x 25 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Iodine, Resublimed (DEA List I Chemical), Crystal, USP is a chemical element and it is often found as a contrast material in X-rays, water purification reagent, nutritional supplement, and as an antiseptic . This product is a List I chemical regulated by The United States Drug Enforcement Agency (DEA) which maintains a list of illicit drugs and the chemical materials used to synthetically produce them. List I chemicals are those that are used in the manufacture of the controlled substances and are important to the manufacture of the substances. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Quantity | 50 kg |
UN Number | UN3495 |
Percent Purity | 99.8 to 100.5% |
Grade | USP |
Packaging | Fiber Drum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction