Learn More
Invitrogen™ Zero Blunt™ PCR Cloning Kit, without competent cells
Offers an easy method for high-efficiency (>80%) cloning of blunt-end PCR products amplified with proofreading, thermostable DNA polymerases
$389.00 - $622.00
Specifications
Bacterial or Yeast Strain | Not Included |
---|---|
Cloning Method | Blunt PCR |
Content And Storage | Zero Blunt™ PCR cloning kits contain linearized pCR™-Blunt vector, ExpressLink™ T4 DNA ligase , 5X ExpressLink™ T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers. Store all components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
Format | Kit |
Product Line | TOPO™, Zero Blunt™ |
Includes: Zero Blunt™ PCR cloning kits contains: linearized pCR™-Blunt vector, ExpressLink™ T4 DNA ligase , 5X ExpressLink™ T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers. Competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.
Description
The Zero Blunt™ PCR Cloning Kit uses the multipurpose cloning vector pCR™-Blunt and ExpressLink™ T4 DNA Ligase to generate a ligation product in a five-minute, room-temperature ligation step.
The pCR-Blunt vector provides:
- EcoR I sites flanking the PCR product insertion site for excision of inserts
- T7 promoter/primer site for in vitro RNA transcription and sequencing
- M13 forward and reverse primer sites for sequencing or PCR screening
The Zero Blunt PCR Cloning Kit is designed to clone blunt PCR fragments (or any blunt DNA fragment) with a low background of non-recombinants. The pCR-Blunt vector contains the lethal E. coliccdB gene fused to the C-terminus of LacZα (Bernard et al., 1994). Ligation of a blunt PCR fragment disrupts expression of the lacZα-ccdB gene fusion permitting growth of only positive recombinants upon transformation. Cells that contain non-recombinant vector are killed when the transformation mixture is plated.
Kit Configurations
The Zero Blunt PCR Cloning Kit is offered in a variety of configurations: with One Shot™ TOPO10 Chemically Competent E. coli (K2700-20 and K2700-40) and without competent cells (K2750-20 and K2750-40) in 20- and 40- reaction kit sizes.
- Fast and convenient: 5-minute, room-temperature ligation
- Efficient: ccdB gene for positive selection results in >80% clones with correct insert
- Flexible: choice of kanamycin or Zeocin™ resistance for flexible antibiotic selection
Cloning, PCR Cloning
Order Info
Shipping Condition: Dry Ice
Specifications
Not Included | |
Zero Blunt™ PCR cloning kits contain linearized pCR™-Blunt vector, ExpressLink™ T4 DNA ligase , 5X ExpressLink™ T4 DNA ligase buffer, control template, dNTPs, sterile water, and M13 forward and reverse primers. Store all components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
|
TOPO™, Zero Blunt™ | |
T7 |
Blunt PCR | |
Kit | |
PCR Cloning Kit | |
Blunt DNA Cloning Vectors |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.